Learn More
Cole-Parmer™ CPC™ Quick Disconnect Sanitary Couplings
Click to view available options
Tubing Size:
1 in.
3/4 in.
Unit Size:
Each
Description
- Attach these couplings directly to your sanitary hardware assemblies
- USP Class IV materials.
- Fully sterilizable.
- Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps.
- Choose from stainless steel or polysulfone inserts to mate to polysulfone bodies.
- All models feature USP Class VI platinum-cured silicone gaskets.
- Mix and match these bodies and inserts with the polysulfone quick-disconnect coupling elements for greater flexibility in your process system.
Specifications
Specifications
| Description | Sanitary x Quick-Disconnect |
| Autoclavable | Autoclavable |
| Material | Polysulfone |
| Tubing Size | 3/4 in. |
| Port Size | 3/4 in. |
| Sterilization Method | Autoclave, e-beam, Ethylene Oxide or Gamma |
| Product Type | Quick-Disconnect, Sanitary Connector |