Learn More
Cole-Parmer™ CPC™ Panel Mount Gasket Couplers
Product Code. p-7138784
Click to view available options
For Use With:
NS4 Series Couplers
NS6 Series Couplers
Unit Size:
Pack of 10
Product Code. 15427333
Brand: Cole-Parmer™ 3160318
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Virtually eliminate any loss of product with this leak-free design
  • Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance.
  • For a complete fitting, order one body and one insert.
  • All bodies will mate with inserts to make a fitting that will work for multiple applications.
Specifications
Type Panel Mount Gasket
Packaging Quantity 10 Gaskets
For Use With NS4 Series Couplers
Material EPDM