Learn More
Cole-Parmer™ CPC™ Panel Mount Gasket Couplers
Click to view available options
For Use With:
NS4 Series Couplers
NS6 Series Couplers
Unit Size:
Pack of 10
Description
- Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance.
- For a complete fitting, order one body and one insert.
- All bodies will mate with inserts to make a fitting that will work for multiple applications.
Specifications
Specifications
| Type | Panel Mount Gasket |
| Packaging Quantity | 10 Gaskets |
| For Use With | NS6 Series Couplers |
| Material | EPDM |