Learn More
Cole-Parmer™ CPC™ DrumQuik PRO System Pro Drum Insert Plug
Product Code. 15467423
Click to view available options
Material:
Polyethylene
Unit Size:
Each
Product Code. 15467423
Brand: Cole-Parmer™ 3160837
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Closed-system design ensures the safety of your plant, the environment and your employees at all phases of chemical handling
  • Easy-to-use, economical solution for chemical dispensing.
  • Durable materials withstand harsh operating conditions.
  • Ideal for 5-gallon jerry cans, 55-gallon drums and IBCs.
  • The DrumQuik PRO system offers a simplified solution to your chemical dispensing needs.
  • Maintains a closed system, protecting your facility, your operating environment, and your employees from the potential dangers of chemical drips, spills and escaping fumes.
  • Simple connections, rugged construction, and chemical-resistant materials handle a wide variety of applications including acids and caustics, agricultural liquids, cleaning agents and sterilants, paints and coatings and food oils and syrups.
  • Base components are a reusable quick-disconnect coupler and a single-use drum insert assembly; both of which are made from environmentally friendly, food-grade polyethylene (PE).
  • Order a convenient dispensing kit which includes all required components, or customize your own system with individual components.
  • Coupler body and plugs mate equally well with all drum inserts listed.
Specifications
Description Buttress Connection
Material Polyethylene
For Use With 5-gallon jerry cans, 55-gallon drums, and IBCs
Flow Rate 10 GPM at less than 2psid (0.14 bar) pressure drop
Maximum Pressure 45 psi
Product Type DrumQuik Pro Drum Insert Plug