Learn More
CP21R7, MedChemExpress
Product Code. 16489496
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
Product Code. 16489496
Brand: Medchem Express HY100207
MedChemExpress CP21R7 is potent GSK-3β inhibitor, with an IC50 of 1.8 nM; CP21R7 also shows inhibitory activitiy against PKCα, with an IC50 of 1900 nM.
CP21R7 is potent GSK-3β inhibitor, with an IC50 of 1.8 nM
Specifications
Chemical Name or Material CP21R7
CAS 125314-13-8
Color Orange
Purity Grade Notes Research
Molecular Formula C19H15N3O2
Quantity 50 mg
Synonym CP21
Solubility Information DMSO : ≥ 32 mg/mL (100.84 mM)
SMILES O=C1C(C2=CN(C)C3=CC=CC=C23)=C(C4=CC(N)=CC=C4)C(N1)=O
Molecular Weight (g/mol) 317.34
Formula Weight 317.34
Percent Purity 98.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.