Learn More
CP-673451, MedChemExpress
Product Code. 16495726
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16495726 100 mg 100mg
16491197 50 mg 50mg
16424958 10 mg 10mg
16417697 5 mg 5mg
4 options
Product Code. 16495726
Brand: Medchem Express HY12050
MedChemExpress CP-673451 is a potent and selective inhibitor of PDGFR with IC50s of 10 and 1 nM for PDGFRα and PDGFRβ, respectively.
CP-673451 is a potent and selective inhibitor of PDGFR with IC50s of 10 and 1 nM for PDGFRα and PDGFRβ, respectively.
Specifications
Chemical Name or Material CP-673451
CAS 343787-29-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H27N5O2
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (239.52 mM)
SMILES NC1CCN(C2=C3N=C(C=CC3=CC=C2)N4C=NC5=CC(OCCOC)=CC=C54)CC1
Molecular Weight (g/mol) 417.5
Formula Weight 417.5
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease