Learn More
COX4NB Mouse anti-Human, Mouse, Rat, Clone: 2B8D3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66547-1-IG-150UL
This item is not returnable.
View return policy
Description
Expressed in liver, pancreas, heart, lung, kidney, brain, skeletal muscle, and placenta, Expression levels are highest in pancreas and moderate in heart, skeletal muscle, and placenta.Specifications
| COX4NB | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43402, O70378, Q5FVL2 | |
| Emc8 | |
| COX4NB Fusion Protein Ag13776 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2B8D3 | |
| Unconjugated | |
| EMC8 | |
| C16orf2, C16orf4, COX4 neighbor, COX4AL, COX4NB, FAM158B, Neighbor of COX4, NOC4, Protein FAM158B | |
| Mouse | |
| Protein G | |
| RUO | |
| 10328, 18117, 361425 | |
| -20°C | |
| Liquid |