missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ COX4I1 (Human) Recombinant Protein
Human COX4I1 full-length ORF ( AAH08704, 1 a.a. - 169 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001327-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLATRVFSLVGKRAISTSVCVRAHESVVKSEDFSLPAYMDRRDHPLPEVAHVKHLSASQKALKEKEKASWSSLSMDEKVELYRIKFKESFAEMNRGSNEWKTVVGGAMFFIGFTALVIMWQKHYVYGPLPQSFDKEWVAKQTKRMLDMKVNPIQGLASKWDYEKNEWKK
Specifications
AAH08704 | |
cytochrome c oxidase subunit IV isoform 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COX4I1 | |
GST |
1327 | |
Wheat germ expression system | |
10 μg | |
COX4, COXIV, MGC72016 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |