missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ COX4 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
Specifications
Specifications
| Gene ID (Entrez) | 1327 |
| Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-39115 |
| Gene Alias | COX IV-1, CO x 4-1, CO x 4 COXIV, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1 cytochrome c oxidase subunit IV, MGC72016 |
| Gene Symbol | COX4I1 |
| Quantity | 0.1 mg |
| Research Category | Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism |
| Storage Buffer | Peptide dissolved in dH2O. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction