Learn More
SureScreen Diagnostics COVID-19 Visual Antigen Rapid Test Cassette
Product Code. 17033987
Change view
Click to view available options
Quantity:
25 Tests/Box
Unit Size:
25 tests
Product Code. Quantity unitSize
17033987 25 Tests/Box 25 tests
1 options
For professional in vitro diagnostic use only.
This product is CE-IVD marked for professional use and complies with the European In-Vitro Diagnostic Devices Directive (98/79/EC).
Product Code. 17033987
Brand: SureScreen Diagnostics COVID19AGVC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For professional in vitro diagnostic use only.
This product is CE-IVD marked for professional use and complies with the European In-Vitro Diagnostic Devices Directive (98/79/EC).
COVID-19 Visual Antigen Rapid Test Cassette using nasal (anterior), nasopharyngeal and oropharyngeal swab samples.
  • For the direct and qualtitative detection of SARS-CoV-2 viral nucleoprotein antigens from nasal (anterior), nasopharyngeal and oropharyngeal swab samples for individuals suspected of COVID-19
  • No additional equipment required
Specifications
CE Marker Yes
Sample Type Nose and Throat Swab
Sensitivity 95%
Type Viral Rapid Test
Quantity 25 Tests/Box
Specificity 100%
Test Time 15 min.