Learn More
Hirschmann™ Cover Glasses for Microscopy
Product Code. 10062481
Change view
Click to view available options
Dimensions (L x W):
15 x 15 mm
18 x 18 mm
20 x 20 mm
22 x 22 mm
22 x 32 mm
22 x 40 mm
22 x 50 mm
22 x 60 mm
24 x 24 mm
24 x 32 mm
24 x 46 mm
24 x 48 mm
24 x 50 mm
Unit Size:
Pack of 1000
Product Code. Dimensions (L x W) unitSize
10062481 18 x 18 mm Pack of 1000
10650193 24 x 24 mm Pack of 1000
13168159 22 x 40 mm Pack of 1000
13128159 22 x 22 mm Pack of 1000
13198159 24 x 32 mm Pack of 1000
13230480 24 x 50 mm Pack of 1000
13158159 22 x 32 mm Pack of 1000
10531942 20 x 20 mm Pack of 1000
13178159 22 x 50 mm Pack of 1000
10165831 15 x 15 mm Pack of 1000
13118169 24 x 46 mm Pack of 1000
13128169 24 x 48 mm Pack of 1000
13188159 22 x 60 mm Pack of 1000
13 options
Product Code. 10062481
Brand: Hirschmann™ 8000102
Only null left
Add to basket
Add to basket
Preserve and protect specimens on slides. Hirschmann Instruments™ Cover Glasses for Microscopy are manufactured from quality glass and come in a range of dimensions.
  • 0.13-0.17 mm thickness
  • Acc. to DIN 58 884, CE acc. IVD 98/79 EC
  • Provided in packs of 1000
  • With lot number on packaging
Specifications
Product Type Coverslip
Thickness (Metric) #1 (0.13 to 0.16 mm)
Dimensions (L x W) 18 x 18 mm
Certifications CE, DIN 58884, IVD 98/79 EC