Learn More
STEMCELL Technologies CoV Protease Substrate-2 EDANS
Product Code. p-7217520
Encompass_Preferred
Click to view available options
Quantity:
100 Tests
1000 Tests
Unit Size:
100 tests
1000 tests
Product Code. 17127641
Brand: STEMCELL Technologies 1000509
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
FRET-based peptide substrate for detection of protease activity of coronaviruses
  • CoV Protease Substrate-2 EDANS allows for efficient detection of coronavirus (CoV) proteases
  • This fluorescence resonance energy transfer (FRET)-based peptide substrate can be cleaved by 3C-like protease (3CLpro), the CoV main protease involved in regulating the coronavirus replication complex
  • 3CLpro is also a potential target for therapeutic applications
  • CoV Protease Substrate-2 EDANS has an excitation wavelength of 336 nm, an emission wavelength of 455 nm, and an extinction coefficient of 5900 cm-1 M-1
  • It consists of a 12 amino acid sequence (VNSTLQSGLRKM) that also contains the DABCYL (quencher) and EDANS (donor) fragments at the N- and C- termini, respectively
  • When the peptide substrate is intact, favorable energetic overlap of the EDANS excited state and the DABCYL absorption leads to efficient intramolecular fluorescence quenching of EDANS by the DABCYL fragment
  • Cleavage of the substrate by CoV proteases, however, dissociates the EDANS fragment from DABCYL, restoring the fluorescence of the EDANS moiety that is directly correlated to the CoV protease activity
Specifications
For Use With (Application) Enzyme Substrate; FRET
Quantity 100 Tests