Learn More
Cole-Parmer™ Straight Fitting, Quick-Disconnect x NPT(M), Acetal
Click to view available options
Tubing Size:
Fits tubing accepting 1/8 in. NPT and quick-disconnect connections
Unit Size:
Pack of 10
Description
- Miniature quick-disconnect couplings are able to handle 1/16 in. ID tubing, and offer a more secure connection than luer fittings.
- Fitting halves engage and seal with a twist.
- Once engaged, the coupling allows the tubing to rotate freely to help prevent kinked tubing and accidental disconnections.
Specifications
Specifications
| Overall Length (Metric) | 2.1844 mm |
| For Use With (Application) | Quick-Disconnect and 1/8 in. NPT thread fitting |
| Tubing Size | Fits tubing accepting 1/8 in. NPT and quick-disconnect connections |