Learn More
Hirschmann™ Counting Chambers
Product Code. 10315421
Change view
Click to view available options
Product Type:
Counting Chambers, Bright lined with clamps; Neubauer-improved
Counting Chambers, Bright lined; Neubauer-improved
Counting Chambers, Neubauer
Counting Chambers, Neubauer-improved
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10315421 Counting Chambers, Bright lined; Neubauer-improved Each
10200872 Counting Chambers, Neubauer Each
10527831 Counting Chambers, Bright lined with clamps; Neubauer-improved Each
10794873 Counting Chambers, Neubauer-improved Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10315421 Supplier Hirschmann™ Supplier No. 8100204
  • ONLINE EXCLUSIVE PRICE
    2005.00 NOK
    1766.52 NOK / Pack of 1
    Save 238.48 NOK
    11% Off
 Restricted Item
Estimated Shipment: 16-02-2027
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann Instruments™ Counting Chambers are available in a variety of depths to fit a range of systems.
  • Double net ruling
  • Available bright-lined or no, with or without clamps
  • Includes 2 cover glasses for haemocytometer
  • CE acc. IVD 98/79 EC
Specifications
Product Type Counting Chambers, Bright lined; Neubauer-improved
Quantity 1Each
Depth (Metric) 0.1 mm
Includes 2 Haemocytometer Glass Covers