Learn More
Corning™ Counting Chamber for Corning Cell Counter
Product Code. 17740789 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
10 μL (approx.) per Chamber
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17740789 10 μL (approx.) per Chamber Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17740789 Supplier Corning™ Supplier No. 480202
Estimated Shipment: 03-11-2026
Disposable counting chambers are composed of two 10 μL chambers. Designed to optimize cell counting throughput by enchaining speed and data accuracy in your day to day mammalian cell culture experiments.
  • Disposable counting chambers are composed of two 10 μL chambers
  • Optimize cell counting throughput by enchaining speed and data accuracy in your day to day mammalian cell culture experiments
TRUSTED_SUSTAINABILITY
Specifications
Product Type Counting Chamber
Capacity (Metric) 10 μL (approx.) per Chamber
Disposable Single-use
Quantity 1/Pk.
Depth (Metric) 0.1 mm
For Use With (Equipment) Corning Cell Counter
Includes 2 Chambers