Learn More
Corning™ Counting Chamber, 0.2 mm
Product Code. 17797014 Shop All Corning Products
Change view
Click to view available options
Depth (Metric):
0.2 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
17797014 0.2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17797014 Supplier Corning™ Supplier No. 480201
  • ONLINE EXCLUSIVE PRICE
    1770.00 NOK
    1601.22 NOK / Pack of 1
    Save 168.78 NOK
    9% Off
Estimated Shipment: 06-04-2027
Reusable glass cell counting slide for use with the Corning™ Cell Counter. This gridless slide has a 0.1 or 0.2 mm depth and comes standard with two high quality cover slips. These slides have a notch for easy loading of cell suspension.
  • Made with reusable glass
  • Gridless
  • 0.2 mm depth
  • Includes two high quality cover slips
  • Feature a notch for easy loading of cell suspension
TRUSTED_SUSTAINABILITY
Specifications
Type Counting Chamber
Material Glass
Volume (Metric) Working 20 μL
Depth (Metric) 0.2 mm
For Use With (Equipment) Corning™ Cell Counter
No. per Pack 1 Pk.