Learn More
Invitrogen™ Countess™ II FL Reusable Slide Holder
Product Code. 15335656 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 unit
Unit Size:
Each
Product Code. Quantity unitSize
15335656 1 unit Each
1 options
Product Code. 15335656
Brand: Invitrogen™ AMEP4746
For use with Countess™ II FL Automated Cell Counter Only. Holds the Countess™ II FL Reusable Slide (A25750). Not compatible with Countess™ disposable slides.
TRUSTED_SUSTAINABILITY
Specifications
Type Reusable Slide Holder
Product Line Countess
For Use With (Application) Cell Counting
For Use With (Equipment) Countess™ Automated Cell Counter
Quantity 1 unit
Shipping Condition Room Temperature