Learn More
Invitrogen™ Countess™ Light Cube Removal Tool
Description
The Countess Light Cube Removal Tool is for use with the Countess II FL and Countess 3 FL automated cell counters. It is used for installation or removal of EVOS Light Cubes from the instruments. The Countess II FL and Countess 3 FL automated cell counters can accommodate up to two Light Cubes simultaneously. With the Countess Light Cube Removal Tool, users can easily remove and install the EVOS Light Cubes, allowing the flexibility to count cells using any of the 15 available fluorescence channels provided by the Light Cubes.
This tool is included with the Countess II FL and 3 FL automated cell counters and can be conveniently stored in the back panel of the instruments when not in use. A replacement Countess Light Cube Removal Tool (Cat. No. AMEP4747) is offered here for separate purchase.
Specifications
Specifications
| Type | Light Cube Remove Tool |
| Product Line | Countess |
| For Use With (Application) | Cell Counting |
| For Use With (Equipment) | Countess II FL and 3 FL Automated Cell Counters |
| Quantity | 1 unit |
| Shipping Condition | Room Temperature |