Learn More
Invitrogen™ Countess™ 3 & 3 FL Power Supply
Product Code. 17816371
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17816371 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17816371 Supplier Invitrogen™ Supplier No. A48207
The Countess 3 and 3 FL Power Supply comes with a power cord and four interchangeable adaptors for global compatibility. It is included with the Countess 3 and 3 FL automated cell counters or a replacement power supply can be purchased separately.
The Countess 3 and 3 FL international power supply is designed for use with the Countess 3 and Countess 3 FL automated cell counters. It should not be used with the original Countess II and Countess II FL automated cell counters. The product includes a power cord and four interchangeable adaptors for United States/Canada/Taiwan/Japan, Europe, and United Kingdom that enable the Countess 3 and 3 FL to be plugged into outlets around the world.
This power supply is included with the Countess 3 and 3 FL automated cell counters (Cat. Nos. AMQAX2000 and AMQAF2000) and is the only recommended power source for use with these instruments. Using other power supplies may damage the Countess 3 and 3 FL counters and affect their performance. A replacement power cord (Cat. No. A48207) is available here for separate purchase.
TRUSTED_SUSTAINABILITY
Specifications
Description Countess II FL, Countess 3 & Countess 3 FL Automated Cell Counters
Compatibility All common global power adaptors
For Use With (Equipment) Countess 3 & 3 FL Automated Cell Counters
Coupler Option Countess Reusable slide
Quantity 1 each