Learn More
Fisherbrand™ Countdown Timer
Use this countdown clock for a variety of timing applications. Fisherbrand™ Countdown Timers are supplied with a Traceable Certificate to ensure accuracy.
115.00 NOK - 167.00 NOK
Specifications
Product Type | Countdown Timer |
---|---|
Alarms | Audible |
Display | Digital |
Quantity | 1 Each |
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
11865311
|
Fisherbrand™
|
60 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11507493
|
Fisherbrand™
|
25 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Timer with full number keyboard
- 76 x 63 x 25mm
- 10 hours
Specifications
Countdown Timer | |
Audible | |
Digital | |
1 Each |