Learn More
Sartorius™ Cotton Linters FN 100 Chromatography Paper
Product Code. 15942720 Shop All Sartorius Products
Change view
Click to view available options
Length (Metric):
190 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15942720 190 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15942720 Supplier Sartorius™ Supplier No. FT15270190100
  • ONLINE EXCLUSIVE PRICE
    4450.00 NOK
    2851.95 NOK / Pack of 1
    Save 1598.05 NOK
    35% Off
Estimated Shipment: 14-12-2026
Specifications
Format Sheet
Grade FN 100
Material Cotton Linters
Width (Metric) 100 mm
Length (Metric) 190 mm
Technique Chromatography