Learn More
Sartorius™ Cotton Linters FN 100 Chromatography Paper
Product Code. 10505141 Shop All Sartorius Products
Change view
Click to view available options
Length (Metric):
200 mm
460 mm
580 mm
Width (Metric):
200 mm
570 mm
600 mm
Description:
100 Sheets
50 Sheets
Unit Size:
Pack of 100
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Description unitSize
10505141 200 mm 200 mm 100 Sheets Pack of 100
10561552 580 mm 600 mm 50 Sheets Pack of 50
10311802 460 mm 570 mm 100 Sheets Pack of 100
10301802 460 mm 570 mm 50 Sheets Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10505141 Supplier Sartorius™ Supplier No. FT2527200200N
Only null left
Specifications
Description 100 Sheets
Format Sheet
Grade FN 100
Material Cotton Linters
Width (English) 200 mm
Width (Metric) 200 mm
Length (Metric) 200 mm
Technique Chromatography