missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Cotton 100, ultra-filtered (UF)

Product Code. 16219841
Change view
Click to view available options
Quantity:
100 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16219841 100 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16219841 Supplier Gibco™ Supplier No. 215381

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cotton 100, UF, is rich in nucleoside content and digested to enhance peptide content.

Cotton 100, UF, is also part of the Peptone Preview Pak which contains:
• Cotton 200, UF—ultra-filtered and rich in nucleoside content, processed to enhance carbohydrate concentrations, which is important for some cell culture and microbial processes
• Wheat 100, UF—an ultra-filtered supplement and good source of glutamine, which makes it ideal for mammalian cell culture applications. It is digested for enhanced peptide content resulting in a low concentration of free amino acids and carbohydrates.
• Soy 100—a highly digested soy protein peptone that is high in nucleosides and free amino acid content. It has high concentrations of vitamins and carbohydrates and is recommend for a wide variety of processes.
• Pea 100, UF—an ultra-filtered peptone digested to enhance nucleoside and free amino acid content. The high total amino acid content and low carbohydrate content are similar to many high-performance animal origin peptones.
• Yeast 100—designed to enhance the content of vitamins and other key nutritional components found in yeast. It is intended for nutritional supplementation for mammalian, microbial, and insect cell culture.

Peptone Preview Pak supplements are non-GMP prototype products that are not for manufacture use. Larger sizes are available through the Gibco Custom Products Services.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Product Type Media Supplement
Quantity 100 g
Source Animal Origin-Free
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.