Learn More
Corning™ Transwell™ Multiple Well Plate with Permeable Polyester Membrane Inserts
Featuring a microscopically transparent polyester membrane that is TC-treated for optimal cell attachment and growth. Corning™ Transwell™ Multiwell Plate with Polyester Membrane Inserts provide better visibility under phase contrast microscopy and allow assessment of cell viability and monolayer formation.
2606.24 NOK - 4866.27 NOK
Specifications
Surface Coating | TC-Treated |
---|---|
Color | Clear |
Surface Treatment | TC-Treated |
Sterility | Sterile |
Product Code | Brand | Pore Size | Description | Culture Area | Diameter (Metric) Outer | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Description | Culture Area | Diameter (Metric) Outer | Price | Quantity & Availability | ||
10317701
|
Corning™
3472 |
3 μm | 12 inserts in a 24-well plate | 0.33 cm2 | 6.5 mm |
4506.83 NOK
Pack of 48 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||
10482181
|
Corning™
3470 |
0.4 μm | 12 inserts in a 24-well plate | 0.33 cm2 | 6.5 mm |
4506.83 NOK
Pack of 48 |
In Stock
Log in to see stock. |
||
13216768
|
Corning™
3464 |
8 μm | 12 inserts in a 24-well plate | 0.33 cm2 | 6.5 mm |
4866.27 NOK
Pack of 48 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||
10357651
|
Corning™
3462 |
3 μm | 12 inserts in a 12-well plate | 1.12 cm2 | 12 mm |
4506.83 NOK
Pack of 48 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||
10565482
|
Corning™
3460 |
0.4 μm | 12 inserts in a 12-well plate | 1.12 cm2 | 12 mm |
4506.83 NOK
Pack of 48 |
In Stock
Log in to see stock. |
||
10619141
|
Corning™
3450 |
0.4 μm | 6 inserts in a 6-well plate | 4.67 cm2 | 24 mm |
2606.24 NOK
Pack of 24 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
10699141
|
Corning™
3452 |
3 μm | 6 inserts in a 6-well plate | 4.67 cm2 | 24 mm |
2606.24 NOK
Pack of 24 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
Description
- Ideal for transport studies, chemotaxis, co-culture, and microbial pathogenesis studies
- Thin membrane and straight pore structure provides consistent transport and cell migration
- Self-centering insert prevents media from wicking between the insert and the outer well
- Plate: Virgin Polystyrene
- Membrane: PET
Specifications
TC-Treated | |
Clear | |
TC-Treated | |
Sterile |