Learn More
Corning™ Snapwell™ Culture Inserts
Product Code. 10555482 Shop All Corning Products
Change view
Click to view available options
Material:
PET
Polycarbonate
Pore Density:
1 x 108 pores/cm2
4 x 106 pores/cm2
Unit Size:
Pack of 24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Pore Density Unit Size
10555482 Polycarbonate 1 x 108 pores/cm2 Pack of 24
10670692 PET 4 x 106 pores/cm2 Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10555482 Supplier Corning™ Supplier No. 3407
  • ONLINE EXCLUSIVE PRICE
    3220.00 NOK
    3038.31 NOK / Pack of 24
    Save 181.69 NOK
    5% Off
In Stock
Designed with a ring supported membrane for use with diffusion or Ussing chambers once cells are grown to confluence. Corning™ Snapwell™ Culture Inserts are primarily used for transport and electrophysiological studies.
The Snapwell insert is a modified Transwell culture insert that contains a 12 mm diameter tissue culture-treated membrane supported by a detachable ring. The inserts are primarily used for transport and electrophysiological studies. Once cells are grown to confluence, this ring-supported membrane can be placed into either vertical or horizontal diffusion or Ussing chambers.
Order Info
Sterile culture inserts are packaged in 6-well cluster plates.
TRUSTED_SUSTAINABILITY
Specifications
Description 6 inserts and 6-well plate
Culture Area 1.12 cm2
Material Polycarbonate
Pore Density 1 x 108 pores/cm2
Pore Size 0.4 μm
Type Culture Insert
Diameter (Metric) Outer 12 mm
No. per Pack 6/Pk.
No. per Case 24/Cs.