Learn More
Corning™ Costar™ Cell Culture Triangular Flasks
GREENER_CHOICE
Product Code. 10466302 Shop All Corning Products
Click to view available options
Closure Color:
Black
Orange
Neck Style:
Angled
Straight
missing translation for 'unitSize'
Pack of 100
Pack of 200
Pack of 500
Product Code. 10466302
missing translation for 'mfr': Corning™ 3275
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from optically clear virgin polystyrene for optimum visibility. Corning™ Cell Culture Triangular Flasks provide excellent accessibility to cell surface.
  • Medical-grade polystyrene is sterile and nonpyrogenic
  • Tissue culture-treated to promote optimal cell attachment and growth
  • Patented canted-neck design improves accessibility for cell monolayer and overall improved handling
  • Graduated
  • Frosted labeling surfaces
  • Extra-wide neck openings for easier pipetting
  • Printed lot number and catalog number on each flask
  • Vented filter caps
  • Maximize gas exchange while minimizing the possibility of contamination
  • Hydrophobic 0.2 μm pore size membrane ensures consistent gas exchange without requiring the cap to be loosened
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Closure Material HDPE
Material Polystyrene
For Use With (Application) For optimal cell attachment and growth
Neck Style Straight
No. per Case 100/Cs.
No. per Pack 5/Pk.
Shape Triangular
Surface Coating Tissue Culture Treated
Graduated Yes
Sterility Sterile
Bottom Shape Modified Triangular
Closure Type Phenolic