Learn More
Corning™ 96-Well Clear PVC Assay Microplates
GREENER_CHOICE
Product Code. 10691432 Shop All Corning Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round
Unit Size:
Pack of 100
Product Code. Well Shape unitSize
10691432 Flat Bottom (F-well) Pack of 100
10574201 Round Pack of 100
10639541 Conical Bottom (V-well) Pack of 100
3 options
Product Code. 10691432
Brand: Corning™ 2595
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from clear flexible Polyvinyl Chloride (PVC) for economy and visibility. Corning™ 96-Well Clear PVC Assay Microplates present an excellent solution for laboratory miniaturization. Due to their flexible nature, these microplates are not compatible with automation.
  • Not treated PVC microplates are economical microplates for solution-based assays, serial dilutions, and general storage applications.
  • Well volume of 250 μL (260 μL for V-bottom); working well volume of 50 μL to 150 μL
  • Lids are not available.
  • Order Info
    Lids are not available. Supplied in packs of 25.
    TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color Clear
    Material PVC
    No. of Wells 96
    Volume (Metric) Working 50 to 150 μL
    Sterility Non-sterile
    Surface Treatment Non-treated
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Solution-based assays, serial dilutions and general storage applications
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.