Learn More
Corning™ Clear Polystyrene 96-Well Microplates
Product Code. 10377601 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10377601 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10377601 Supplier Corning™ Supplier No. 3366
  • ONLINE EXCLUSIVE PRICE
    4510.00 NOK
    4210.83 NOK / Pack of 100
    Save 299.17 NOK
    6% Off
Estimated Shipment: 14-04-2027
Add to basket
Add to basket
Bulk Quote Request
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
TRUSTED_SUSTAINABILITY
Specifications
Binding Property High Binding
Lid Without Lid
Array 8 x 12
Packaging Bags of 25
Binding Type High Bind
Color Clear
Product Type EIA/RIA
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Surface Treatment High Binding
Well Shape Round
For Use With (Application) Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions.
No. per Case 100/Cs.
No. per Pack 25/Pk.
Quantity 100/Cs.