Learn More
Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates
Product Code. 10363173 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Natural
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 25
Pack of 50
Product Code. Color Barcode unitSize
10363173 Clear Includes Barcode Pack of 50
10727811 Natural No Barcode Pack of 25
2 options
Product Code. 10363173
Brand: Corning™ 3758
Only null left
Add to basket
Add to basket
Constructed of medical grade polypropylene to accommodate wide temperature changes that might occur from storage to thermal cycling. Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.
Corning Thermowell GOLD 96-well PCR microplates are offered in four formats to ensure maximum flexibility and a perfect match for your applications. The original Thermowell 96-well PCR microplates are universal fit and can be cut into 3 x 8 well segments.
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Certified DNase and RNase-free
Format Standard
Description 2 generic bar code labels per plate
Array 8 x 12
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Total Volume (Metric) Well 200 μL
Well Shape Conical Bottom (V-well)
For Use With (Application) Reverse transcription (RT)-PCR
Includes 2 generic bar code labels per plate
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Pk.