missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Coronin 7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559668
This item is not returnable.
View return policy
Description
Immunogen sequence: TRVFLYELLP ESPFFLECNS FTSPDPHKGL VLLPKTECDV REVELMRCLR LRQSSLEPVA FRLPRVRKEF FQDDVFPDTA VIWEPVLSAE AWLQGANGQP W Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 80%.
Coronins, such as CORO7, constitute an evolutionarily conserved family of WD-repeat actin-binding proteins. CORO7 plays a role in Golgi complex morphology and function (Rybakin et al., 2004, 2006 [PubMed 15327992] [PubMed 16905771]).
Specifications
| Coronin 7 | |
| Polyclonal | |
| Unconjugated | |
| CORO7 | |
| 0610011B16Rik; 70 kDa WD repeat tumor rejection antigen; 70 kDa WD repeat tumor rejection antigen homolog; AW048373; AW556392; CORO7; coronin 7; coronin-7; CRN7; POD1; tumor specific antigen 70 kDa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79585 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57737 | |
| CORO7 | |
| Recombinant protein corresponding to Human CORO7-PAM16. Recombinant protein control fragment (Product #RP-98663). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction