Learn More
CORO6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17243-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| CORO6 | |
| Polyclonal | |
| Unconjugated | |
| CORO6 | |
| CORO6, coronin 6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 216961, 245982, 84940 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6QEF8, Q920J3, Q920M5 | |
| CORO6 | |
| CORO6 Fusion Protein Ag11167 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |