missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CORO1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17760-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Alternative splicing results in multiple transcript variants. A related pseudogene has been defined on chromosome 16.Specifications
| CORO1A | |
| Polyclonal | |
| Unconjugated | |
| CORO1A | |
| CLABP, Clipin A, CLIPINA, CORO1, CORO1A, Coronin 1A, Coronin like protein A, Coronin like protein p57, HCORO1, p57, TACO | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11151, 12721, 155151 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O89053, P31146, Q91ZN1 | |
| CORO1A | |
| CORO1A Fusion Protein Ag12139 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction