missing translation for 'onlineSavingsMsg'
Learn More

CORO1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16857724 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16857724 20 μL 20µL
16847724 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16857724

Brand: Proteintech 177601AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Alternative splicing results in multiple transcript variants. A related pseudogene has been defined on chromosome 16.

Specifications

Antigen CORO1A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CORO1A
Gene Accession No. O89053, P31146, Q91ZN1
Gene Alias CLABP, Clipin A, CLIPINA, CORO1, CORO1A, Coronin 1A, Coronin like protein A, Coronin like protein p57, HCORO1, p57, TACO
Gene Symbols CORO1A
Host Species Rabbit
Immunogen CORO1A Fusion Protein Ag12139
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11151, 12721, 155151
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.