Learn More
Corning™ Thermowell™ GOLD Polypropylene PCR Tubes
GREENER_CHOICE
Product Code. 10473952 Shop All Corning Products
Click to view available options
Color:
Assorted
Clear
Closure Type:
Domed Cap
Flat Cap
Volume (Metric):
0.2 mL
0.5 mL
Unit Size:
Pack of 1000
Product Code. 10473952
Brand: Corning™ 3747
Designed for precise fit in heat blocks
Individual PCR tubes are made of thin wall polypropylene and designed for precise fit in heat blocks to optimize heat transfer. Tubes are RNase-/DNase-free, are autoclavable at 121°C, and withstand centrifugation to 10,000 x g.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase-free
Closure Color Clear
Color Clear
Volume (Metric) 0.2 mL
Quantity 500/Pk.
Closure Material Polypropylene
For Use With (Equipment) Heat blocks to optimize heat transfer
Shape Flat
Sterility Non-sterile
Material Polypropylene
Max. Force 10,000 x g
Closure Type Domed Cap
Autoclavable Autoclavable
Format Tube
Product Type Thin Wall PCR Tube