Learn More
Corning™ Phenolic GPI Screw Caps for Disposable Culture Tubes
GREENER_CHOICE
Product Code. 10602624
Change view
Click to view available options
Closure Size:
13-415
15-415
18-415
Height (Metric):
13.5 mm
16 mm
17.5 mm
Unit Size:
Pack of 1000
Pack of 250
Product Code. Closure Size Height (Metric) unitSize
10602624 18-415 17.5 mm Pack of 1000
10426843 13-415 13.5 mm Pack of 1000
11359263 13-415 13.5 mm Pack of 1000
11952348 18-415 17.5 mm Pack of 250
10150403 15-415 16 mm Pack of 1000
5 options
Product Code. 10602624
Brand: Corning™ 9999918
Only null left
Add to basket
Add to basket
With white rubber liner
Made of a phenolic compound. These caps have a white rubber liner.
TRUSTED_SUSTAINABILITY
Specifications
Closure Size 18-415
Liner Material Rubber
Product Type Screw Cap
Autoclavable Autoclavable
Material Phenolic
Color Black
Height (Metric) 17.5 mm
No. per Pack 1000/Pk.
No. per Case 1000/Cs.
Tip Material Rubber
For Use With (Equipment) Culture Tubes