Learn More
Corning™ Robolids with Corner Notch
GREENER_CHOICE
Product Code. 10702783 Shop All Corning Products
Change view
Click to view available options
Format:
384-Well Robolid
96-Well Robolid
Unit Size:
Pack of 50
Product Code. Format unitSize
10702783 96-Well Robolid Pack of 50
10702973 384-Well Robolid Pack of 50
2 options
Product Code. 10702783
Brand: Corning™ 3090
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with automation equipment
  • Combines the sealing ability of a storage mat with the rigidity of a plastic lid
  • Designed for repeated application and removal by automation and for preventing short-term evaporation
  • Silicone sealing plugs for organic solvent resistance and low extractables
  • Can be used manually or with automation
  • Order Info
    Available in 96- and 384-well formats.
    TRUSTED_SUSTAINABILITY
    Specifications
    Sterility Non-sterile
    No. of Wells 96
    No. per Case 50/Cs.
    No. per Pack 25/Pk.
    Format 96-Well Robolid