Learn More
Corning™ Reusable Plastic Reagent Bottles with GL-45 PP Screw Cap
Shop All Corning Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
Unit Size:
Pack of 10
Pack of 20
Description
These bottles are manufactured from translucent polypropylene. They are wide-mouth and supplied with PP screw top caps.
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Translucent |
| Material | Polypropylene |
| No. per Case | 20/Cs. |
| No. per Pack | 1/Pk. |
| Sterility | Non-sterile |
| Shape | Cylindrical |
| Height (Metric) | 13.2 cm |
| Diameter (Metric) | 7.3 cm |