Learn More
Corning™ Reusable Plastic Erlenmeyer Flasks with Red Graduations
Product Code. p-4901379 Shop All Corning Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
Graduations:
10 mL
20 mL
50 mL
100 mL
200 mL
Unit Size:
Pack of 4
Pack of 6
Product Code. 17223554
Brand: Corning™ 4990P50
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Translucent polymethylpentene; with polypropylene screw cap
Translucent polymethylpentene flasks are manufactured with red graduations for reference. The flasks have a screw neck and are supplied with a PP cap.
TRUSTED_SUSTAINABILITY
Specifications
Closure Material Polypropylene
Capacity (Metric) 50 mL
Material Polypropylene
Graduations 10 mL
Neck Style Narrow Mouth
No. per Case 6/Cs.
No. per Pack 1/Pk.
Closure Type Screw Cap