Learn More
Corning™ PureCoat™ Fibronectin Peptide Cell Culture Flasks with Vented Cap
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
Brand:  Corning™ 356242
Additional Details : Weight : 1.21500kg
 View more versions of this product
Product Code. 15350001
  • 6250.00 NOK / Pack of 10
Estimated Shipment: 02-07-2024
Description
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Specifications
Specifications
75 cm2
Canted
5/Pk.
PureCoat E cm Mimetic Fibronectin Peptid
Polystyrene
10/Cs.
Rectangular
Vented