Learn More
Corning™ PTFE-Coated Stir Bars
Product Code. 10327673 Shop All Corning Products
Change view
Click to view available options
Length (Metric):
25.4 mm
Length (English):
1 in.
For Use With (Equipment):
PC-200 Hot Plates/Stirrers
Unit Size:
Each
Product Code. Length (Metric) Length (English) For Use With (Equipment) unitSize
10327673 25.4 mm 1 in. PC-200 Hot Plates/Stirrers Each
1 options
Product Code. 10327673
Brand: Corning™ 409021
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for use with Scholar™ PC-171 stirrers
  • PTFE-coated for increased chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) 0.31 in.
For Use With (Equipment) PC-200 Hot Plates/Stirrers
Length (English) 1 in.
Diameter (Metric) 8 mm
Length (Metric) 25.4 mm
Material Magnet
Quantity 1