Learn More
Corning™ PTFE-Coated Magnetic Stir Bars
GREENER_CHOICE
Product Code. 10626953 Shop All Corning Products
Change view
Click to view available options
Length (Metric):
25.4 mm
50.8 mm
76.2 mm
Diameter (Metric):
9.5 mm
19 mm
Length (English):
1 in.
2 in.
3 in.
Diameter (English):
0.37 in.
0.75 in.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Length (English) Diameter (English) unitSize
10626953 76.2 mm 19 mm 3 in. 0.75 in. Each
10183943 50.8 mm 9.5 mm 2 in. 0.37 in. Each
10628653 25.4 mm 9.5 mm 1 in. 0.37 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10626953 Supplier Corning™ Supplier No. 409000
For use in Corning Hotplate Stirrers
Corning magnetic stir bars are polytetrafluorethylene (PTFE)-coated for increased chemical ­resistance. 
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) 0.75 in.
For Use With (Equipment) PC-611 hotplate stirrers
Length (English) 3 in.
Coating PTFE
Diameter (Metric) 19 mm
Length (Metric) 76.2 mm
Material Magnet
Quantity 1
Type Stir Bar