Learn More
Corning™ Osteo Assay Surface Microplates
GREENER_CHOICE
Product Code. 11397791 Shop All Corning Products
Click to view available options
Volume (Metric) Working:
380 to 570 μL
75 to 200 μL
Packaging:
1/Pk.; 2/Cs.
1/Pk; 4/Cs
Unit Size:
Pack of 2
Pack of 4
Product Code. 11397791
Brand: Corning™ 3989
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Capable of supporting the functional properties of osteogenic cells. Corning™ Osteo Assay Surface delivers lot-to-lot consistency, translating to reliable and reproducible results in bone cell assays. The surface also offers a consistent and defined alternative to preparing dentine or bone slices, reducing the variability in assay systems and resulting in more predictable assay readouts.
  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Array 8 x 12
Packaging 1/Pk.; 2/Cs.
Color Clear
Product Type 96-well Osteo Assay Surface Multiwell Microplate
Material Polystyrene
Volume (Metric) Well 360 μL
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Osteo Assay
For Use With (Application) Osteoblast and osteoclast differentiation and functional studies
No. per Case 2/Cs.
No. per Pack 1/Pk.
Well Color Clear