Learn More
Corning™ LSE™ Digital Dry Bath Accessory, Single Blocks
Shop All Corning Products
Click to view available options
No. of Places:
5
6
12
20
24
Tube Size:
0.5 mL
10 mm
12 mm
13 mm
1.5 mL
16 mm
2 mL
25 mm
15 mL
50 mL
Unit Size:
Each
Description
The single and dual block configurations are used in the Corning LSE Digital Dry Bath Heaters.
Specifications
Specifications
| For Use With (Equipment) | LSE Dry Bath Heater |
| Material | Stainless Steel |
| No. of Places | 20 |
| Type | Block |
| Tube Size | 10 mm |