Learn More
Corning™ DNA-BIND™ 96-Well Plates
Product Code. 10691072 Shop All Corning Products
Click to view available options
Color:
Clear
White
Quantity:
10/Cs.
50/Cs.
Unit Size:
Pack of 10
Pack of 50
Product Code. 10691072
Brand: Corning™ 2499
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Convenient method for covalently immobilizing ssDNA by either the 5' or 3' end
  • Ready to use without surface preparation, prehybridization, or addition of expensive crosslinking reagents or highly purified oligos
  • Ideal for hybridization and amplification
  • Surface provides binding with high specificity and extremely low background
  • Flat-bottom wells
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color White
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment DNA-Bind
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Ideal for hybridization and amplification
Quantity 50/Cs.
Well Color White