Learn More
Corning™ DeckWorks™ Low Binding Hinged Rack Barrier Tips
Product Code. 11333665 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 100 μL
1 to 20 μL
1 to 200 μL
Unit Size:
Pack of 3840
Pack of 960
Product Code. 11333665
Brand: Corning™ 4138
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures
Corning DeckWorks low binding barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1 to 200 μL
Filtered Filtered
Sterility Sterile
Color Natural
For Use With (Application) Used for quantitative assays in which sample binding can significantly alter results
Format Hinged Rack
Tip Style Low Retention, Round
Packaging Rack
Graduated Yes
Product Line DeckWorks
Material HDPE