Description
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resalable polyethylene bags. Tip sizes include 10 μL, 200 μL, and 1000 μL and offer maximum savings.
Specifications
Specifications
| Volume (Metric) | 100 to 2000 μL |
| Sterility | Non-sterile |
| Color | Natural |
| For Use With (Application) | Help reduce the production and waste of plastic materials |
| Tip Style | Round |
| Disposable | Single-use |
| For Use With (Equipment) | DeckWorks Racks |
| Graduated | Yes |
| Product Line | DeckWorks |
| Material | Polypropylene |