Description
96-well Microplates
- Features 96 large reservoir (reagent) wells and 96 corresponding protein wells
- Conical bottom protein wells allow improved centering of the protein drop.
- Compatible with manual pipettors and automation
- Novel merged well design provides efficient vapor space for protein crystallization.
- Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
384-well Microplate
- Meets ANSI/SBS standard footprint dimensions for microplates
- Ideal for fully automated crystal screening
- Features 192 reservoir wells and 192 corresponding protein wells
- Flat bottom protein wells are optimized for imaging of crystals.
- Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
Specifications
Specifications
| Binding Property | Hydrophilic |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | Clear |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | High Binding |
| Well Shape | Flat Bottom (F-well) |
| Includes | 96 flat-bottom reservoir wells and 96 conical-bottom protein wells |
| Show More |