Learn More
Corning™ Cryogenics Workstation
GREENER_CHOICE
Product Code. 10725912 Shop All Corning Products
Click to view available options
Type:
Cryogenic Vial Rack
Cryogenic Workstation
Unit Size:
Each
Product Code. 10725912
Brand: Corning™ 430526
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rack grips Corning externally and internally threaded self-standing vials, so caps can be removed with one hand
  • Reusable polycarbonate rack and storage tray
  • Removable rack has an alphanumeric grid and holds 30 vials
  • Tray can also be used to hold water or crushed ice
  • Rack and tray can be autoclaved at 121°C (250°F)
Order Info
Workstation dimensions: 14.63L x 5.69W x 3 in. H (37.1 x 14.5 x 7.6cm)
TRUSTED_SUSTAINABILITY
Specifications
Color White
Holds 30 Cryogenic Vials
Material Polycarbonate
Type Cryogenic Vial Rack