Learn More
Corning™ Collagen IV, Mouse
Product Code. 10376931 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 mg
10 x 1 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10376931 1 mg 1mg
10602322 10 x 1 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10376931 Supplier Corning™ Supplier No. 354233
  • ONLINE EXCLUSIVE PRICE
    2160.00 NOK
    2126.23 NOK / 1mg
    Save 33.77 NOK
    1% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 to 5 μg/cm2
For Use With (Application) Cell Attachment and Proliferation
Formulation Frozen in 0.05 M Hydrochloric Acid (HCL)
Quantity 1 mg
Product Type Collagen, Mouse Type IV
Content And Storage -70°C