Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
Specifications
Specifications
| Concentration | 5 μg/cm2 |
| For Use With (Application) | Cell Attachment and Proliferation, Cell-specific Morphology and Function |
| Formulation | Liquid in 15 mm acetic acid |
| Quantity | 5 mg |
| Source | Bovine Articular Cartilage |
| Product Type | Collagen, Bovine Type II |
| Content And Storage | -70°C |
| Purity or Quality Grade | >90% by SDS-PAGE |